missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CADM4 Polyclonal Antibody
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
Specifications
Specifications
| Antigen | CADM4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | CADM4 |
| Gene Accession No. | Q8NFZ8 |
| Gene Alias | Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor |
| Gene Symbols | CADM4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?