missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-8 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98949AF350
This item is not returnable.
View return policy
Description
Cadherin-8 Polyclonal specifically detects Cadherin-8 in Human samples. It is validated for ELISA.
Specifications
| Cadherin-8 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1006 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 350 | |
| cadherin 8, type 2, cadherin-8, Nbla04261, putative protein product of Nbla04261 | |
| Produced in rabbits immunized with purified, recombinant Cadherin-8 (NP_001787.2; Met1-Met621). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction