Learn More
Cadherin-23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-59271
Product Code. 18777883
  • 483.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Cadherin-23 Polyclonal specifically detects Cadherin-23 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cadherin related 23, cadherin-23, cadherin-like 23, cadherin-related 23, cadherin-related family member 23, CDHR23, DKFZp434P2350, FLJ00233, FLJ36499, KIAA1774, KIAA1812, MGC102761, Otocadherin, USH1D
Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit
IgG
CDH23
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.