Learn More
Cadherin-23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59271
This item is not returnable.
View return policy
Description
Cadherin-23 Polyclonal specifically detects Cadherin-23 in Human samples. It is validated for Western Blot.
Specifications
| cadherin related 23, cadherin-23, cadherin-like 23, cadherin-related 23, cadherin-related family member 23, CDHR23, DKFZp434P2350, FLJ00233, FLJ36499, KIAA1774, KIAA1812, MGC102761, Otocadherin, USH1D | |
| Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
| IgG |
| CDH23 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |