missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-13 Antibody (392411), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3264PEATT594
This item is not returnable.
View return policy
Description
Cadherin-13 Monoclonal antibody specifically detects Cadherin-13 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| Cadherin-13 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 392411 | |
| Flow Cytometry | |
| cadherin 13, H-cadherin (heart), cadherin-13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, Truncated cadherin, truncated-cadherin | |
| Mouse myeloma cell line NS0-derived recombinant human Cadherin-13, Glu23-Ala692, Accession # P55290 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 1012 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction