missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CACNB3 Antibody, Novus Biologicals™

Product Code. 30067191 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30067191 25 μg 25µg
30066806 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30067191 Supplier Novus Biologicals™ Supplier No. NBP33083225ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CACNB3 Polyclonal antibody specifically detects CACNB3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 5-10 μg/mL
Formulation Phosphate buffered saline
Gene Alias CAB3, CACNLB3, Calcium channel voltage-dependent subunit beta 3, calcium channel, voltage-dependent, beta 3 subunit, voltage-dependent L-type calcium channel subunit beta-3
Host Species Rabbit
Immunogen Synthetic peptide derived from the rat CACNB3 conjugated to KLH
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 784
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.