missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNA1E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30954-25ug
This item is not returnable.
View return policy
Description
CACNA1E Polyclonal antibody specifically detects CACNA1E in Human samples. It is validated for Western Blot
Specifications
| CACNA1E | |
| Polyclonal | |
| Western Blot : 1:400 | |
| Brain calcium channel II, CACH6calcium channel, R type, alpha-1 polypeptide, CACNL1A6calcium channel, voltage-dependent, alpha 1E subunit, calcium channel, voltage-dependent, R type, alpha 1E subunit, Cav2.3, L type, alpha-1 polypeptide, L type, alpha-1 polypeptide, isoform 6, voltage-dependent calcium channel alpha 1E subunit, voltage-dependent R-type calcium channel subunit alpha-1E, voltage-gated calcium channel alpha 1E subunit, Voltage-gated calcium channel subunit alpha Cav2.3 | |
| Synthetic peptide derived from the rat CACNA1E conjugated to KLH | |
| 25 μg | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 777 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction