Learn More
Fisherbrand™ Cable for Electrode S7 or S8 Head
Product Code. 11756398 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
DIN
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
11756398 DIN Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11756398
Please to purchase this item. Need a web account? Register with us today!
Cable accessory for electrodes with S7 or S8 head is ideal for measuring pH in many lab and scientific environments.
  • Coaxial connector for DIN plug 19262
  • For combined electrodes
  • S7 Cable DIN connector, 3M
  • Knick, WTW, Polymetron, Schott, Sidel
Specifications
Connector Types DIN
For Use With (Equipment) Electrodes with S7 or S8 head
Type Cable accessory
Length (Metric) Cable 3 m