Learn More
CA125/MUC16 Antibody (10F10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26127-100ul
 View more versions of this product
Product Code. 30037660
  • 533.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CA125/MUC16
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
10F10
ELISA, Immunohistochemistry 1:50-1:200
CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
A synthesized peptide derived from Human CA125/MUC16 [UniProt Q8WXI7]
100 μL
Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers
94025
Store at -20 to -70C. Avoid freeze-thaw cycles.