missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C9orf64 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15919514
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15919514 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15919514 Supplier Invitrogen™ Supplier No. PA566828

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SDDLLKKLLKG EMLSYGDRQE VEIRGCSLWC VELIRDCLLE LIEQKGEKPN GEINSILLDY YLWDYAHDHR EDMKGIPFHR IRCIY Highest antigen sequence identity to the following orthologs - mouse 79%, rat 74%.

Involved in salvaging queuosine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf64
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C9orf64
Gene Accession No. Q5T6V5
Gene Alias C9orf64; chromosome 9 open reading frame 64; Queuosine salvage protein; UPF0553 protein; UPF0553 protein C9orf64
Gene Symbols C9orf64
Host Species Rabbit
Immunogen Recombinant Human C9orf64. Recombinant protein control fragment (Product #RP-105386).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.