missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C8orf55 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13434359
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13434359 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13434359 Supplier Invitrogen™ Supplier No. PA532685

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Hamster, Opossum, Zebrafish (89%) Panda, Mosquito, Ant (83%).

DSCD75 is a 208 amino acid membrane protein with one transmembrane helix. Current testing suggests that the cDNA of DSCD75 may be expressed by osteogenic human mesenchymal stem cells. DSCD75 may be involved in some transporter activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C8orf55
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene THEM6
Gene Accession No. Q8WUY1
Gene Alias 4930572J05Rik; C13H8orf55; C14H8orf55; C8orf55; DSCD75; mesenchymal stem cell protein DSCD75; mesenchymal stem cell protein DSCD75 homolog; protein THEM6; PSEC0098; RGD1359378; Them6; Thioesterase superfamily member 6; UPF0670 protein C8orf55; UPF0670 protein C8orf55 homolog; UPF0670 protein THEM6; UPF0670 protein THEM6 homolog
Gene Symbols THEM6
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human C8orf55.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51337
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.