Learn More
C7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17642-1-AP-150UL
This item is not returnable.
View return policy
Description
C7 is a component of the complement system. It participates in the formation of Membrane Attack Complex (MAC). People with C7 deficiency are prone to bacterial infection.Specifications
| C7 | |
| Polyclonal | |
| Unconjugated | |
| C7 | |
| C7, complement component 7, Complement component C7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109828, 117517, 730 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10643 | |
| C7 | |
| C7 Fusion Protein Ag11819 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |