missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C6orf58 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16360275
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16360275 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16360275 Supplier Invitrogen™ Supplier No. PA583317

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GTSNLSETEP PLWKESPGQL SDYRVENSMY IINPWVYLER MGMYKIILNQ TARYFAKFAP DNEQNILWGL PLQYGWQYRT GRLADPTRR Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 64%.

C6orf58 is a protein coding gene. May be involved in early liver development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf58
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C6orf58
Gene Accession No. Q6P5S2
Gene Alias C6orf58; chromosome 6 open reading frame 58; LEG1; Protein LEG1 homolog; UPF0762 protein C6orf58
Gene Symbols C6orf58
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LEG1. Recombinant protein control fragment (Product #RP-95022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 352999
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.