missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1q Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00000712-D01
This item is not returnable.
View return policy
Description
C1q Polyclonal antibody specifically detects C1q in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| C1q | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| A chain, complement component 1, q subcomponent, A chain, complement component 1, q subcomponent, alpha polypeptide | |
| C1QA (NP_057075.1, 1 a.a. - 245 a.a.) full-length human protein. MEGPRGWLVLCVLAISLASMVTEDLCRAPDGKKGEAGRPGRRGRPGLKGEQGEPGAPGIRTGIQGLKGDQGEPGPSGNPGKVGYPGPSGPLGARGIPGIKGTKGSPGNIKDQPRPAFSAIRRNPPMGGNVVIFDTVITNQEEPYQNHSGRFVCTVPGYYYFTFQVLSQWEICLSIVSSSRGQVRRSLGFCDTTNKGLFQVVSGGMVLQLQQGDQVWVEKDPKKGHIYQGSEADSVFSGFLIFPSA | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 712 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| No additive | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction