missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C19orf55 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15741352 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15741352 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15741352 Supplier Invitrogen™ Supplier No. PA563578

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PTAVNVTSAS HAVAPLQEIK QNLHTWNSSL LDLETLSLQS RAARLLKRSK ASIS Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 61%.

PROSER3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf55
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PROSER3
Gene Accession No. Q2NL68
Gene Alias C10H19orf55; C19orf55; C1H19orf55; LOW QUALITY PROTEIN: proline and serine-rich protein 3; proline and serine rich 3; proline and serine-rich protein 3; PROSER3; RGD1565712; uncharacterized protein C19orf55 homolog
Gene Symbols PROSER3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf55. Recombinant protein control fragment (Product #RP-101674).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.