missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C19orf12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144268
This item is not returnable.
View return policy
Description
C19orf12 Polyclonal Antibody for ELISA, ICC/IF, IHC (P)
Specifications
| C19orf12 | |
| Polyclonal | |
| Unconjugated | |
| C19orf12 | |
| C19orf12; chromosome 19 open reading frame 12; NBIA3; NBIA4; neurodegeneration with brain iron accumulation 3; protein C19orf12; SPG43 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 83636 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9NSK7 | |
| C19orf12 | |
| Recombinant Human Protein C19orf12 protein (65-104AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction