missing translation for 'onlineSavingsMsg'
Learn More

C16orf57 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene

Product Code. 17505254
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17505254 30 μL 30µL
17585244 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17505254 Supplier Origene Technologies Supplier No. TA811328S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C16orf57 Monoclonal antibody specifically detects C16orf57 in Bacteria samples. It is validated for Western Blot

Specifications

Antigen C16orf57
Applications Western Blot
Classification Monoclonal
Clone OTI1B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias AA960436; C16orf57; FLJ13154; HAMAP-Rule:MF_03040}; hUsb1; HVSL motif containing 1; HVSL1; Mpn1; mutated in poikiloderma with neutropenia protein 1; PN; putative U6 snRNA phosphodiesterase; RGD1305215; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA biogenesis phosphodiesterase 1; U6 snRNA phosphodiesterase; U6 snRNA phosphodiesterase {ECO:0000255; UPF0406 protein C16orf57; UPF0406 protein C16orf57 homolog; USB1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-1166 of human CAS9 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.