missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ c-Cbl Monoclonal Antibody (1762CT401.80.60)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537728
This item is not returnable.
View return policy
Description
c-Cbl Monoclonal Antibody for Western Blot
Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
Specifications
| c-Cbl | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P22681 | |
| Cbl | |
| KLH conjugated synthetic peptide between 450-644 amino acids from human CBL. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| 1762CT401.80.60 | |
| Unconjugated | |
| Cbl | |
| 4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL | |
| Mouse | |
| Protein G | |
| RUO | |
| 867 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction