missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-C1 Heavy Chain Receptor Binding Domain Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5425-SP
This item is not returnable.
View return policy
Description
BoNT-C1 Heavy Chain Polyclonal specifically detects BoNT-C1 Heavy Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| BoNT-C1 Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BoNTC1 Heavy Chain, DcD | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10523893 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P18640 | |
| CbC4_4096 | |
| E. coli-derived recombinant BoNT-C1 Heavy Chain Asn866-Glu1291 Accession # P18640 | |
| 25 μg | |
| Primary | |
| Detects C. botulinum BoNT-C1 Heavy Chainin direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant (r) BoNT-D-HCR is observed and 5% cross-reactivity with rBoNT-A-HCR, rBoNT-A-LC, and rBoNT-B-LC is observed. | |
| C. botulinum | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction