missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxytoluene, Carl Roth™

Product Code. 10735164
Encompass
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging
10735164 250 g Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10735164 Supplier Carl Roth Supplier No. 3559.1
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 128-37-0
Melting Point 69°C to 71°C
Packaging Plastic
Molecular Formula C15H24O
Quantity 250 g
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Molecular Weight (g/mol) 220.36
PubChem CID 31404
ChEBI CHEBI:34247
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.