missing translation for 'onlineSavingsMsg'
Learn More

Burixafor hydrobromide, MedChemExpress

Product Code. 16470408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470408 5 mg 5mg
16451296 200 mg 200mg
16437948 10 mg 10mg
16431906 500 mg 500mg
16427746 100 mg 100mg
16413887 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16470408 Supplier Medchem Express Supplier No. HY19867A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Burixafor hydrobromide (TG-0054 hydrobromide) is an orally bioavailable and potent antagonist of CXCR4 and a well anti-angiogenic drug that is of potential value in treating choroid neovascularization. Burixafor hydrobromide (TG-0054 hydrobromide) mobilizes mesenchymal stem cells, attenuates inflammation, and preserves cardiac systolic function in a porcine model of myocardial infarction.

Burixafor hydrobromide (TG-0054 hydrobromide) is an orally bioavailable and potent antagonist of CXCR4 and a well anti-angiogenic drug that is of potential value in treating choroid neovascularization. Burixafor hydrobromide (TG-0054 hydrobromide) mobilizes mesenchymal stem cells, attenuates inflammation, and preserves cardiac systolic function in a porcine model of myocardial infarction.

Specifications

Chemical Name or Material Burixafor hydrobromide
Physical Form Solid
CAS 1191450-19-7
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H51N8O3P.5/2HBr
For Use With (Application) COVID-19-immunoregulation
Synonym TG-0054 hydrobromide
Solubility Information H2O : 50 mg/mL (65.02 mM; Need ultrasonic)
SMILES NC1=CC(N2CCN(CCP(O)(O)=O)CC2)=NC(NC[C@H]3CC[C@H](CNCCCNC4CCCCC4)CC3)=N1.[H]Br.[5/2]
Molecular Weight (g/mol) 769.01
Formula Weight 769.01
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.