missing translation for 'onlineSavingsMsg'
Learn More

Bumetrizole-d3, TRC

Product Code. 17978689
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978689 2.5 mg 2.50mg
17958779 1 mg 1mg
17947979 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17978689 Supplier Toronto Research Chemicals Supplier No. TRCB6896822.5MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C17 D3 H15 Cl N3 O
Molecular Weight (g/mol) 318.82
Synonym 2-(5-Chloro-2H-benzotriazol-2-yl)-6-(1,1-dimethylethyl)-4-methylphenol-d3, 2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-p-cresol-d3, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chloro-2H-benzotriazole-d3, 5-Chloro-2-(3-tert-butyl-2-hydroxy-5-methylphenyl)benzotriazole-d3, Uvinul 3026-d3, VS 550-d3, Viosorb 550-d3;
IUPAC Name 2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-(trideuteriomethyl)phenol
SMILES [2H]C([2H])([2H])c1cc(c(O)c(c1)C(C)(C)C)n2nc3ccc(Cl)cc3n2

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H18ClN3O/c1-10-7-12(17(2,3)4)16(22)15(8-10)21-19-13-6-5-11(18)9-14(13)20-21/h5-9,22H,1-4H3/i1D3
Formula Weight 318.1327
Percent Purity >95%
Chemical Name or Material Bumetrizole methyl-d3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.