Learn More
BUCHI Multivapor Frit Set
Product Code. 10493665 Shop All Buchi Products
Click to view available options
For Use With (Equipment):
P-6/P-12 Multivapor
This item is not returnable. View return policy
Product Code. 10493665
Brand: BUCHI 044856
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Specifications
Material Polyethylene
Type Porous Frit Set
For Use With (Application) Silica adsorption and foam protection
For Use With (Equipment) P-6/P-12 Multivapor