Learn More
- Home
- Products
- Evaporators
- Evaporator Glassware Sets
- BUCHI Evaporating Flasks for Rotary Evaporators
BUCHI Evaporating Flasks for Rotary Evaporators
Product Code. 12024592 Shop All Buchi Products
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Taper:
24/40
29.2/32
29.2/42
Unit Size:
Each
Material (Coating):
Plastic and Glass
Uncoated
21 product options available for selection
| Product Code. | Volume (Metric) | Taper | unitSize | Material (Coating) |
|---|---|---|---|---|
| 12024592 | 5000 mL | 29.2/32 | Each | - |
| 12831494 | 100 mL | 29.2/32 | Each | Uncoated |
| 10406021 | 250 mL | 29.2/32 | Each | Uncoated |
| 10280592 | 500 mL | 29.2/32 | Each | Uncoated |
| 12861494 | 1000 mL | 29.2/32 | Each | Uncoated |
| 10466201 | 2000 mL | 29.2/32 | Each | Uncoated |
| 10551172 | 3000 mL | 29.2/32 | Each | Uncoated |
| 12704963 | 500 mL | 29.2/42 | Each | Uncoated |
| 12784963 | 1000 mL | 29.2/42 | Each | Uncoated |
| 12734973 | 2000 mL | 29.2/42 | Each | Uncoated |
| 12744973 | 3000 mL | 29.2/42 | Each | Uncoated |
| 10648662 | 1000 mL | 29.2/32 | Each | Plastic and Glass |
| 10496941 | 500 mL | 29.2/32 | Each | Plastic and Glass |
| 10128741 | 2000 mL | 29.2/32 | Each | Plastic and Glass |
| 12755113 | 3000 mL | 29.2/32 | Each | Plastic and Glass |
| 10352182 | 250 mL | 29.2/32 | Each | Plastic and Glass |
| 12778833 | 100 mL | 29.2/32 | Each | Plastic and Glass |
| 12788833 | 50 mL | 29.2/32 | Each | Plastic and Glass |
| 10178831 | 50 mL | 29.2/32 | Each | Uncoated |
| 10257813 | 1000 mL | - | Each | - |
| 10033503 | 2000 mL | 24/40 | Each | Uncoated |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- High-performance flasks for rotary evaporators
- Exclusive use of DURAN borosilicate glass 3.3
- High chemical resistance against acids, alkalis and organic substances
- Resistant to thermal shocks and high temperatures combined with low thermal expansion
- Constant monitoring of glass wall thickness uniformity
- High degree of attention paid to sphericity of rotating glass parts
- Glass parts are tension-relieved at 560°C
- Available in volumes from 50 mL to 3000 mL
- Available uncoated or with P+G coating
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Type | Evaporating Flask |
| For Use With (Equipment) | Rotavapor |
| Includes | w/o Plastic and glass coating |
| Taper | 29.2/32 |
| Volume (English) | 169.07 oz. |
| Volume (Metric) | 5000 mL |