missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20514-1-AP-150UL
This item is not returnable.
View return policy
Description
Shows marked antiproliferative activity, being able to induce G(1) arrest.Specifications
| BTG4 | |
| Polyclonal | |
| Unconjugated | |
| BTG4 | |
| B cell translocation gene 4, BTG family member 4, BTG4, PC3B, Protein BTG4, Protein PC3b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315650, 54766, 56057 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70552, Q9NY30 | |
| Btg4 | |
| BTG4 Fusion Protein Ag14339 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction