missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BTF Polyclonal antibody specifically detects BTF in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | BTF |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 330-552 of human BTF (NP_001070909.1).,, Sequence:, REEEFKTHHEMKEYSGFAGVSRPRGTFFRIRGRGRARGVFAGTNTGPNNSNTTFQKRPKEEEWDPEYTPKSKKYFLHDDRDDGVDYWAKRGRGRGTFQRGRGRFNFKKSGSSPKWTHDKYQGDGIVEDEEETMENNEEKKDRRKEEKE |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?