missing translation for 'onlineSavingsMsg'
Learn More

BTBD3 Polyclonal Antibody, Invitrogen™

Product Code. 15747691 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15747691 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15747691 Supplier Invitrogen Supplier No. PA559842

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAADIFPRKK PANSSSTSVQ QYHQQNLSNN NLIPAPNWQG LYPTIRERNA MMFNNDL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

BTBD3 is a protein containing BTB/POZ domain. Its function has not been determined yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD3
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BTBD3
Gene Accession No. P58545, Q9Y2F9
Gene Alias BTB (POZ) domain containing 3; BTB domain containing 3; BTB/POZ domain containing protein 3; BTB/POZ domain-containing protein 3; BTBD3; dJ742J24.1; KIAA0952; mKIAA0952
Gene Symbols BTBD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BTBD3
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228662, 22903
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.