Learn More
Paragon Scientific™ BS/U-tube Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
- Supplied with full UKAS ISO 17025 calibration
Specifications
Specifications
| Viscosity Range | 0.9 to 3 cSt |
| Calibrated | Yes |
| Approximate Constant | 0.003 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Includes | UKAS ISO 17025 calibration |
| Size | A |
8 week lead-time