Learn More
Paragon Scientific™ BS/U-tube Viscometer for Transparent Liquids
Product Code. p-7161828
Click to view available options
Viscosity Range:
0.9 to 3 cSt
2.0 to 10 cSt
20 to 100 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Unit Size:
Each
Product Code. 16080450
Brand: Paragon Scientific™ U01NC
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 0.9 to 3 cSt
Calibrated No
Approximate Constant 0.003
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Size A
4 week lead-time