Learn More
Paragon Scientific™ BS/U-tube Viscometer for Transparent Liquids
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
| Viscosity Range | 20 to 100 cSt |
| Calibrated | No |
| Approximate Constant | 0.1 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | D |
4 week lead-time