missing translation for 'onlineSavingsMsg'
Learn More

BS-181 hydrochloride, MedChemExpress

Product Code. 16411777
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16411777

missing translation for 'mfr': Medchem Express HY13266A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BS-181 hydrochloride is a highly selective CDK7 inhibitor with IC50 of 21 nM, and > 40-fold selective for CDK7 than CDK1, 2, 4, 5, 6, or 9.

BS-181 hydrochloride is a highly selective CDK7 inhibitor with IC50 of 21 nM, and > 40-fold selective for CDK7 than CDK1, 2, 4, 5, 6, or 9.

Specifications

Chemical Name or Material BS-181 hydrochloride
Physical Form Solid
CAS 1397219-81-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H33ClN6
For Use With (Application) Cancer-Kinase/protease
Solubility Information H2O : 100 mg/mL (239.81 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (119.91 mM)
SMILES CC(C1=C(N=C(C=C2NCC3=CC=CC=C3)NCCCCCCN)N2N=C1)C.[H]Cl
Molecular Weight (g/mol) 416.99
Formula Weight 416.99
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.