missing translation for 'onlineSavingsMsg'
Learn More

Bromosuccinic acid, 96%

Product Code. p-7026684
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11469857

Brand: Thermo Scientific Alfa Aesar B22829.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 923-06-8 | C4H5BrO4 | 196.98 g/mol

Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

Solubility
Insoluble in ether.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923-06-8
Molecular Formula C4H5BrO4
Molecular Weight (g/mol) 196.98
MDL Number MFCD00004213
InChI Key QQWGVQWAEANRTK-UHFFFAOYNA-N
Synonym bromosuccinic acid, 2-bromosuccinic acid, monobromosuccinic acid, butanedioic acid, bromo, +--bromosuccinic acid, dl-bromosuccinic acid, rs-bromosuccinic acid, succinic acid, bromo, bromobutanedioic acid
PubChem CID 73557
ChEBI CHEBI:73712
IUPAC Name 2-bromobutanedioic acid
SMILES OC(=O)CC(Br)C(O)=O

Specifications

Melting Point 164°C to 166°C (decomposition)
Quantity 5 g
Beilstein 1723556
Merck Index 14,1437
Solubility Information Insoluble in ether.
Formula Weight 196.99
Percent Purity 96%
Chemical Name or Material Bromosuccinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.