Learn More
Bromo-Peg3-Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | Bromo-Peg3-Bromide |
| CAS | 31255-10-4 |
| Molecular Formula | C6H12O2Br2 |
| Quantity | 10 mg |
| Synonym | Ethane, 1,2-Bis(2-Bromoethoxy)- |
| InChI Formula | InChI=1S/C6H12Br2O2/c7-1-3-9-5-6-10-4-2-8/h1-6H2 |
| SMILES | BrCCOCCOCCBr |
| IUPAC Name | 1,2-bis(2-bromoethoxy)ethane |
| Molecular Weight (g/mol) | 275.96 |
| Formula Weight | 273.9204 |