Learn More
Brivanib (alaninate), MedChemExpress
Product Code. 16494866
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494866 100 mg 100mg
16493828 10 mg 10mg
16456557 5 mg 5mg
16430207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16494866 Supplier Medchem Express Supplier No. HY10336
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Brivanib alaninate (BMS-582664) is an ATP-competitive inhibitor against VEGFR2 with an IC50 of 25 nM; has moderate potency against VEGFR-1 and FGFR-1, but more than 240-fold against PDGFRβ.
Brivanib alaninate (BMS-582664) is an ATP-competitive inhibitor against VEGFR2 with an IC50 of 25 nM
Specifications
Chemical Name or Material Brivanib (alaninate)
CAS 649735-63-7
Color White
Purity Grade Notes Research
Molecular Formula C22H24FN5O4
Quantity 100 mg
Synonym BMS-582664
Solubility Information DMSO : 100 mg/mL (226.52 mM; Need ultrasonic)
SMILES O=C(O[C@@H](COC1=CN2C(C(OC3=C(C4=C(NC(C)=C4)C=C3)F)=NC=N2)=C1C)C)[C@@H](N)C
Molecular Weight (g/mol) 441.46
Formula Weight 441.46
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease