missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Brilliant Blue G 250

Product Code. 10168371
Encompass
Change view
Click to view available options
Quantity:
10 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10168371 10 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10168371 Supplier Carl Roth™ Supplier No. 9598.1
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Blue
Form Liquid
For Use With (Application) Protein Analysis in Gel Electrophoresis
Molecular Weight (g/mol) 854.03g/mol
Quantity 10 g
Product Type Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.