Learn More
MedChemExpress Bremelanotide Acetate
Product Code. 16453597
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453597 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16453597 Supplier MedChemExpress Supplier No. HY18678A
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
MedChemExpress Bremelanotide Acetate (PT-141 Acetate), a synthetic peptide analogue of α-MSH, is an agonist at melanocortin receptors including the MC3R and MC4R for the treatment of sexual dysfunction.
Specifications
Purity 98.18%
CAS 1607799-13-2
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 1085.22 g/mol
Product Type Peptides
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 36 mg/mL (33.17 mM)
Synonym PT-141 Acetate