missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD4 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00 € - 483.00 €
Specifications
| Antigen | BRD4 |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18311414
|
Novus Biologicals
NBP3-15953-20UL |
20 μg |
201.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18387604
|
Novus Biologicals
NBP3-15953-100UL |
100 μg |
483.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BRD4 Polyclonal antibody specifically detects BRD4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDownSpecifications
| BRD4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 23476 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1 | |
| A synthetic peptide corresponding to a sequence within amino acids 623-722 of human BRD4 (NP_055114.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title