missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA2, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31069.100uL
This item is not returnable.
View return policy
Description
Sequence: VHPISLSSSKCHDSVVSMFKIENHNDKTVSEKNNKCQLILQNNIEMTTGTFVEEITENYKRNTENEDNKYTAASRNSHNLEFDGSDSSKNDTVCIHKDETDLLFTDQHNICLKLSGQFMKEGNTQIKEDLSSpecifications
| BRCA2 | |
| Polyclonal | |
| Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| breast cancer 2, early onset | |
| BRCA2 | |
| Recombinant protein corresponding to human BRCA2. | |
| RUO | |
| 675 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunofluorescence | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| BRCC2/FACD/FAD/FAD1/FANCB/FANCD/FANCD1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction