missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BRCA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17270024 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17270024 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17270024 Supplier Invitrogen™ Supplier No. PA5117528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BRCA2
Gene Accession No. 0
Gene Alias brca 2; BRCA1/BRCA2-containing complex, subunit 2; BRCA2; brca-2; BRCA2 DNA repair associated; BRCA2, DNA repair associated; BRCC2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; GLM3; mutant BRCA2; PNCA2; RAB163; RP11-298P3.4; XRCC11
Gene Symbols BRCA2
Host Species Rabbit
Immunogen Recombinant Zebrafish Breast cancer 2, early onset protein (2737-2874aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 566758
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.