missing translation for 'onlineSavingsMsg'
Learn More

Branebrutinib, MedChemExpress

Product Code. 16424588
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424588

Brand: Medchem Express HY112161

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Branebrutinib (BMS-986195) is a highly potent, selective covalent, irreversible inhibitor of Bruton’s tyrosine kinase (BTK), with an IC50 of 0.1 nM.

Branebrutinib (BMS-986195) is a highly potent, selective covalent, irreversible inhibitor of Bruton’s tyrosine kinase (BTK), with an IC50 of 0.1 nM.

Specifications

Chemical Name or Material Branebrutinib
CAS 1912445-55-6
Color White
Purity Grade Notes Research
Molecular Formula C20H23FN4O2
Quantity 10 mg
Synonym BMS-986195
Solubility Information DMSO : ≥ 100 mg/mL (269.96 mM)
SMILES O=C(C1=CC(F)=C(N2C[C@@H](NC(C#CC)=O)CCC2)C3=C1NC(C)=C3C)N
Molecular Weight (g/mol) 370.42
Formula Weight 370.42
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.