missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ Polypropylene Screw Caps

Product Code. 15292829
Click to view available options
Thread Size (Metric):
GL 18
GL 32
GL 45
Unit Size:
Pack of 12
Pack of 24
This item is not returnable. View return policy

Product Code. 15292829

Brand: BRANDTECH™ VITLAB™ 83312

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Screw caps for VITLAB™ and other GL threaded bottles

  • Polypropylene construction
  • Transparent
  • Autoclavable at 121°C

Specifications

Color Transparent
Material Polypropylene
Sterility Non-sterile
Thread Size (Metric) GL 32
Disposable Limited-use
Product Type Screw Cap
For Use With (Equipment) Vitlab and other GL threaded bottle
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.