Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Inlet Separators
Click to view available options
For Use With:
RE2, RZ2, RE 2.5, RZ 2.5
RE5, RZ5, RE6, RZ6, RE 2.5, RZ 2.5
RE8, RE9, RE16, RZ8, RZ9, RZ16
Unit Size:
Each
Description
- For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
- Inlet Separators 'AK' help keep condensates and particulates from entering the vacuum pump, preserving performance.
Note: For RE/RZ 2.5: For rotary vane pumps RE 2.5 (article no. 2079715x) and RZ 2.5 (article no. 2079812x) - please order Inlet Separator 20698006. For rotary vane pumps RE 2.5 (article no. 2069715x) and RZ 2.5 (article no. 2069812x) - please order Inlet Separator 20698000.
Specifications
Specifications
| For Use With (Application) | To keep condensates and particulates from entering the vacuum pump |
| For Use With | RE5, RZ5, RE6, RZ6, RE 2.5, RZ 2.5 |
| Model | 20698006 |
| Type | Inlet Catchpot/Separator |