Learn More
BRAND™ Dispensette™ Bottletop Dispenser Discharge Tube
Product Code. 10681182 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.5, 1, 2, 5, 10 mL
25, 50, 100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10681182 0.5, 1, 2, 5, 10 mL Each
10519212 25, 50, 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10681182 Supplier BRAND™ Supplier No. 707935
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with the Dispensette III™ and Organic dispensers, for dispensing aggressive and organic solvents, respectively.
  • With integrated valve
  • Provided individually
  • Fine tip shape
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Fine tip
Type Replacement Discharge Tube
Volume (Metric) 0.5, 1, 2, 5, 10 mL
For Use With (Equipment) 0.5, 1, 2, 5, 10mL Fine tip Dispensette Organic
Quantity 1/Pk.
Length (Metric) 90 mm