missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Globally Harmonized System (GHS) Flammable Picto Labels

Product Code. p-4569857
Click to view available options
Quantity:
39/Pk.
Material:
Vinyl
Height (English):
0.62 in.
Width (English):
0.6 in.
Unit Size:
Pack of 39
This item is not returnable. View return policy

Product Code. 15640351

Brand: Brady™ 118821

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For flammable gases, liquids and solids; pyrophorics; self-reatives; self-heating

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.

  • Legend: Flammable
  • Made of Vinyl or Coated paper
  • Available in three sizes

Specifications

Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Adhesive Type Permanent Acrylic
Color Black, Red, White
Finish Gloss
Format Label
Material Vinyl
Product Type Pictogram Label
Sterility Non-sterile
Display Text Flammable
For Use With (Application) Flammable gases, liquids and solids
Quantity 39/Pk.
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Dimensions (LxW) 0.625 x 0.625 in.
Width (English) 0.6 in.
Height (English) 0.62 in.
Height (Metric) 15.20 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.