Learn More
Brady™ BMP™ 71 PermaSleeve™ Wire Marker Sleeves
Description
- Has 3:1 shrink ratio
- Heat shrinkable
- Fade resistant
- Flame retardant
- Made of B-342 Permasleeve PS polyolefin material
Specifications
Specifications
| Certifications/Compliance | SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202, RoHS Compliant, UL Recognized; Tested per: ASTM E162 and ASTM E662 |
| Description | Bulk BMP71 PermaSleeve Wire Marker Sleeves |
| Material | Polyolefin (B-342) |
| Product Type | Wire Marker Sleeve |
| For Use With (Application) | Wire and cable marking, Panel identification |
| For Use With (Equipment) | 22 to 16 wire ga |
| Quantity | 1 |
| Length (Metric) | 15.24 m |
| Diameter (English) | 0.187 to 0.450 in. |
| Diameter (Metric) | 4.75 to 11.43 mm |