Learn More
BPIPP, TRC
Specifications
Specifications
| Chemical Name or Material | BPIPP |
| CAS | 325746-94-9 |
| Molecular Formula | C22H16BrN3O3 |
| Quantity | 1 mg |
| Synonym | 5-(3-Bromophenyl)-5,11-dihydro-1,3-dimethyl-1H-indeno[2',1':5,6]pyrido[2,3-d]pyrimidine-2,4,6(3H)-trione |
| InChI Formula | InChI=1S/C22H16BrN3O3/c1-25-20-17(21(28)26(2)22(25)29)15(11-6-5-7-12(23)10-11)16-18(24-20)13-8-3-4-9-14(13)19(16)27/h3-10,15,24H,1-2H3 |
| SMILES | O=C1N(C)C2=C(C(C3=CC=CC(Br)=C3)C(C(C4=C5C=CC=C4)=O)=C5N2)C(N1C)=O |
| IUPAC Name | 5-(3-bromophenyl)-1,3-dimethyl-5,11-dihydro-1H-indeno[2',1':5,6]pyrido[2,3-d]pyrimidine-2,4,6(3H)-trione |
| Molecular Weight (g/mol) | 450.28 |
| Formula Weight | 449.04 |