Learn More
Gibco™ Bovine Fetuin (custom product)
Product Code. 10505053
Click to view available options
Quantity:
1 g
Unit Size:
1g
For Research Use Only. All usage must comply with product instructions.
Product Code. 10505053
Brand: Gibco™ 10344026
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Bovine Fetuin (custom product)
This is a GIBCO Custom Product. Please complete and submit the Custom Media Inquiry Form and a Customer Service Representative will contact you regarding pricing and availability.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2 to 8°C
Description Bovine Fetuin, 1g, Used for Cell Viability, Proliferation and Function, Mammalian Cell Culture Applications, For Research Use Only
Product Type Bovine Fetuin
For Use With (Application) Cell viability, proliferation and function, mammalian cell culture
Quantity 1 g
Regulatory Status For research use only