Learn More
Stuart™ Bottle Top Adapters
Click to view available options
Thread Size:
38 mm
40 mm
Unit Size:
Each
Description
- Available in various thread sizes
- Polypropylene
- 1 Adapter/Cs.
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Equipment) | Liquid Handling Products |
| Quantity | 1 Adapter/Cs. |
| Thread Size | 40 mm |