Learn More
Karl Hecht™ Bottle, Narrow Neck with Glass Stopper
Product Code. 10575952
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
Color:
Amber
Clear
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
10575952 500 mL Amber Each
10409981 250 mL Amber Each
10185661 50 mL Clear Each
10287471 250 mL Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10575952 Supplier Karl Hecht™ Supplier No. 41303024
Only null left
Add to basket
Add to basket
Round-shouldered chemical/reagent bottle. Karl Hecht™ Bottle, Narrow Neck with Glass Stopper includes a ground-in flat glass stopper.
  • Narrow neck.
  • Round shoulder.
  • Ground in flat glass stopper.
  • Available in clear glass or amber glass.
Specifications
Closure Material Glass
Color Amber
Capacity (Metric) 500 mL
Material Glass
Neck Style Narrow
Product Type Bottle
Shape Round Shoulder
Closure Type Stopper