Learn More
Sampling Systems™ Bottle Liquid Sampler
Product Code. 15889321 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
1000 mm
1500 mm
Includes:
500 mL Glass Bottle
500 mL Glass Bottle, Bottle Cover
Unit Size:
Each
Product Code. 15889321
Brand: Sampling Systems™ 5031S1500
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Incorporates a spring loaded mechanism, which means that liquid only flows in when the handle is squeezed allowing samples to be taken from set locations. Sampling Systems™ Bottle Liquid Sampler takes samples straight into the bottle.
  • 316L Stainless Steel sampler body
  • Glass bottle
  • Single point
  • Sampling for low to medium viscosity liquids
  • Ideal for low volume (unit dose) sampling
  • The sampler is available with a cover that stops the bottle getting wet
Specifications
Product Type Liquid Sampler
Material Stainless Steel with Glass Bottle
Includes 500 mL Glass Bottle
Capacity (Metric) 500 mL
Length (Metric) 1500 mm