Learn More
BRAND™ Bottle Adapters for Dispensette™ and seripettor™ Bottletop Dispensers and Titrette™ Bottletop Burettes
Product Code. 10727411 Shop All Brand Products
Change view
Click to view available options
Thread Size:
External Thread: 33 mm; Bottle Thread: 38 mm
External Thread: 33 mm; Bottle Thread: 40 mm
External Thread: 33 mm; Bottle Thread: 45 mm
Material:
PTFE, ETFE
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Material unitSize
10727411 External Thread: 33 mm; Bottle Thread: 45 mm PTFE, ETFE Each
10170331 External Thread: 33 mm; Bottle Thread: 38 mm PTFE, ETFE Each
10449652 External Thread: 33 mm; Bottle Thread: 40 mm PTFE, ETFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10727411 Supplier BRAND™ Supplier No. 704395
Only null left
Add to basket
Add to basket
Adapters for common bottle threads and standard taper joints
  • Economical polypropylene adapters work for most applications
  • ETFE/PTFE adapters for aggressive acids and solvents
  • Leading number refers to male thread of adapter
  • Second number refers to bottle neck the adapter fits
  • 28 mm, 24 mm and 22 mm bottle threads require stacking adapter with 45/33 mm adapter.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE, ETFE
Neck Type Bottle Thread: 45 mm
Thread Size External Thread: 33 mm; Bottle Thread: 45 mm
Diameter (Metric) Neck 45 mm
Diameter (Metric) Outer Neck 33 mm